Learn More
d,l-Metamfepramone.HCl, Lipomed
Product Code. 30462857
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462857 Supplier Lipomed Supplier No. LPMMAF1548HC50
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
d,l-Metamfepramone.HCl
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,N-Dimethylcathinone (hydrochloride)
CAS 10105-90-5
Analyte or Component Names N,N-Dimethylcathinone (hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C11 H15 N O . Cl H
Synonym Dimethylcathinone Hydrochloride, N,N-Dimethylcathinone Hydrochloride, Metamfepramone Hydrochloride, 1-Propanone, 2-(dimethylamino)-1-phenyl-, hydrochloride (1:1), 1-Propanone, 2-(dimethylamino)-1-phenyl-, hydrochloride (9 CI), Propiophenone, 2-(dimethylamino)-, hydrochloride (6 CI,7 CI,8 CI), Propiophenone, 2-(dimethylamino)-, hydrochloride (6 CI,7 CI,8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CC(N(C)C)C(=O)c1ccccc1
IUPAC Name 2-(dimethylamino)-1-phenylpropan-1-one;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 213.7
Quantity 50 mg
Formula Weight 213.092 g/mol
Show More Show Less
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.