missing translation for 'onlineSavingsMsg'
Learn More

D-Histidine, free acid, MP Biomedicals™

Product Code. 11413831 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413831 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413831 Supplier MP Biomedicals Supplier No. 0215126380

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 351-50-8
Color White
Molecular Formula C6H9N3O2
MDL Number MFCD00065963
Synonym d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
InChI Key HNDVDQJCIGZPNO-RXMQYKEDSA-N
SMILES N[C@H](CC1=CN=CN1)C(O)=O
IUPAC Name (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular Weight (g/mol) 155.16
Quantity 100 g
PubChem CID 71083
ChEBI CHEBI:27947
Formula Weight 155.15
Physical Form Free-flowing Powder
Chemical Name or Material D-Histidine, free acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.