missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Gulono-γ-Lactone, 99.6%, For HPLC analysis, MP Biomedicals™

Product Code. 11453751 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Product Code. Quantity unitSize
11453751 10 g 10g
11443751 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11453751

Brand: MP Biomedicals 0215121710

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6322-07-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
InChI Key SXZYCXMUPBBULW-UHFFFAOYNA-N
Synonym d-gulono-1,4-lactone, d---gulonic acid gamma-lactone, gulonolactone, d-gulonic acid gamma-lactone, gulono-1,4-lactone, d-gulonic acid-1,4-lactone, 3r,4s,5s-5-r-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, 3r,4s,5s-5-1r-1,2-dihydroxyethyl-3,4-dihydroxyoxolan-2-one, d-gulono-gamma-lactone, gulonic acid-1,4-lactone
PubChem CID 165105
IUPAC Name (3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Color White
Assay Percent Range 99.6%
Quantity 10 g
Formula Weight 178.1
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Gulono-γ-Lactone, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.