missing translation for 'onlineSavingsMsg'
Learn More

α-D-Glucose Pentaacetate, MP Biomedicals™

Product Code. 11443150 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443150 100 g 100g
11433150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443150 Supplier MP Biomedicals Supplier No. 0210047080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Quantity 100 g
Formula Weight 390.3
Physical Form Solid
Chemical Name or Material α-D-Glucose Pentaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.