Learn More
D-Glucose-6-phosphate dipotassium salt hydrate, ≥95%, MP Biomedicals™
Product Code. 11403000 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403000 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11403000 Supplier MP Biomedicals Supplier No. 0210031301
  • ONLINE EXCLUSIVE PRICE
    176.00 EUR
    166.95 EUR / 1g
    Save 9.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Specifications
CAS 5996-17-8
Color White
Assay Percent Range ≥95%
Molecular Formula C6H13K2O9P
Quantity 1 g
Synonym d-glucose 6-phosphate dipotassium*hydrate sigma g
InChI Key LJSYGYHQPZYSEZ-FAOVPRGRSA-N
SMILES C(C(C(C(C(C=O)O)O)O)O)OP(=O)(O)O.[K].[K]
IUPAC Name potassium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] dihydrogen phosphate
Molecular Weight (g/mol) 338.331
PubChem CID 131855786
Formula Weight 390.4
Physical Form Crystalline Powder
Chemical Name or Material D-Glucose-6-phosphate dipotassium salt hydrate
Show More Show Less
compliance-icons
Product Identifier
  • D-Glucose-6-phosphate Dipotassium Salt Hydrate
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 2
Hazard Statement
  • H371-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: D-glucose-6-phosphate dipotassium salt hydrate