missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Galactosamine hydrochloride, 99%, MP Biomedicals™

Product Code. 11448090 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448090 5 g 5g
11438090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448090 Supplier MP Biomedicals Supplier No. 0210174305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1772-03-8
Molecular Formula C6H13NO5·HCl
Synonym Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactopyranose hydrochloride

Specifications

Color White
Assay Percent Range 0.99
Quantity 5 g
Formula Weight 215.64
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Galactosamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.