CAS RN 1772-03-8
CAS RN 1772-03-8
IUPAC Name:
hydrogen (3R,4R,5R,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol chloride
Synonyms:
hydrogen galactosamine chloride
Molecular Weight (g/mol):
215.63
Molecular Formula:
C6H14ClNO5
InChi Key:
QKPLRMLTKYXDST-BMZZJELJSA-N
SMILES:
[H+].[Cl-].N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
1
–
4
of
4
results
1
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 1772-03-8 Molecular Formula: C6H13NO5·HCl MDL Number: MFCD00135830 Synonym: Chondrosamine hydrochloride,2-Amino-2-deoxy-D-galactose hydrochloride
2
D-(+)-Galactosamine hydrochloride, 99%, MP Biomedicals™
CAS: 1772-03-8 Molecular Formula: C6H13NO5·HCl Synonym: Chondrosamine hydrochloride,2-Amino-2-deoxy-D-galactopyranose hydrochloride
3
D-Galactosamine, Hydrochloride, TRC
CAS: 1772-03-8 Molecular Formula: C6 H13 N O5 . Cl H Molecular Weight (g/mol): 215.63 IUPAC Name: (2R,3R,4R,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride SMILES: Cl.N[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)CO