missing translation for 'onlineSavingsMsg'
Learn More

D-Arabinitol-1,2-13D2, TRC

Product Code. 30334656
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334656 5 mg 5mg
30340582 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30334656 Supplier Toronto Research Chemicals Supplier No. TRCA7356055MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Arabinitol-1,2-13C2
Molecular Formula 13C2 C3 H12 O5
Quantity 5 mg
Synonym D-Arabitol-1,2-13C2, D-Lyxitol-1,2-13C2
InChI Formula InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m1/s1/i1+1,3+1/t3-,4-,5+
SMILES O[13CH2][13C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2R,3R,4R)-(4,5-13C2)pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 154.131
Formula Weight 154.075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.