missing translation for 'onlineSavingsMsg'
Learn More

d-Aminolevulinic Acid Hydrochloride, MP Biomedicals™

Product Code. 11408292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408292 Supplier MP Biomedicals Supplier No. 0219462180
  • ONLINE EXCLUSIVE PRICE
    65.05 EUR
    61.95 EUR / 100mg
    Save 3.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

d-Aminolevulinic acid, formed from glycine and succinyl CoA, by ALA synthase, is the first compound in prophyrin biosynthesis pathway. It serves as an intermediate in heme biosynthesis.

Specifications

Melting Point 150°C to 160°C
CAS 9-2-5451
Molecular Formula C5H10ClNO3
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
Solubility Information Soluble in water (5% v/w-clear, colorless solution).
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
SMILES C(CC(=O)O)C(=O)CN.Cl
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
Molecular Weight (g/mol) 167.59
Quantity 100 mg
PubChem CID 123608
Formula Weight 167.59g/mol
Percent Purity ∼98%
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.