Learn More
D-Alanine tert-butyl ester hydrochloride, 98%
Product Code. 15476338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476338 1 g 1g
15486338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15476338 Supplier Thermo Scientific Alfa Aesar Supplier No. H59898.03
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
CAS: 59531-86-1 | C7H16ClNO2 | 181.66 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59531-86-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00077087
InChI Key WIQIWPPQGWGVHD-NUBCRITNSA-N
Synonym d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 16211919
IUPAC Name tert-butyl (2R)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OC(C)(C)C)N.Cl
Specifications
Beilstein 5763077
Quantity 1 g
Formula Weight 181.66
Optical Rotation −1.4° (c=2 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material D-Alanine tert-butyl ester hydrochloride