Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Product Code. 11996716
Change view
Click to view available options
Volume (Metric):
30 μL
300 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1920
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Unit Size
11996716 300 μL Non-sterile Pack of 1920
10660404 300 μL Sterile Pack of 1920
10121343 30 μL Non-sterile Pack of 1920
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11996716 Supplier Thermo Scientific™ Supplier No. 551611
  • ONLINE EXCLUSIVE PRICE
    257.00 EUR
    245.00 EUR / Pack of 1920
    Save 12.00 EUR
    4% Off
Estimated Shipment: 01-04-2027
Includes
20 magazines per case
D.A.R.T.'S (Disposable Automation Research Tips) have been designed for the ultimate performance in automated pipetting
Unlike conventional automation tips, D.A.R.T. tips are provided in disposable rigid magazines that seal onto the pipetting heads by sealing against a silicone pad. This forms a definitive seal which minimizes tip-to-tip height variations and elimiates static interactions, enabling consistent dispensing into the bottom of microplate wells.
  • Compatible with the Matrix Hydra DT and Matrix PlateMate 2x3 and PlateMate Plus Systems
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision
  • Thorough inspection for proper formation as well as straightness ensures tips are free of defects
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced, eliminating extra handling of the tips which can cause damage or contamination
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 300 μL
Sterility Non-sterile
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Thermo Scientific Versette, Thermo Scientific Matrix PlateMate 2x3, Thermo Scientific Matrix Hydra DT
Product Line D.A.R.T.S