Learn More
Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal antibody specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Classification | Polyclonal |
| Content And Storage | Store at 4C in the dark. |