Learn More
Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS
Product Code. 11398359 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398359 100 mg 100mg
11308369 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11398359 Supplier MP Biomedicals Supplier No. 02100071.1
  • ONLINE EXCLUSIVE PRICE
    69.45 EUR
    66.15 EUR / 100mg
    Save 3.30 EUR
    4% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Cytosine-β-D-Arabinofuranoside Hydrochloride
Application Notes:
  • It is used as an antineoplastic and antiviral agent.
Usage Statement:
  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
  • Selective inhibitor of DNA synthesis
Specifications
Product Type Nucleoside
Quantity 100 mg
Color White
Molecular Formula C9H14ClN3O5
CAS 69-74-9