missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 15 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunofluorescence applications with Epredia™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.

133.00 EUR - 571.20 EUR

Specifications

Antigen Cytokeratin 15 Ab-1
Clone LHK15
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Concentration Quantity Price Quantity & Availability  
Product Code Brand Concentration Quantity Price Quantity & Availability  
12618826
View Documents Promotion Details
Epredia™
MS-1068-P1
200μg/mL 500 μL
364.00 EUR
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12608826
View Documents Promotion Details
Epredia™
MS-1068-P0
200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12698816
View Documents Promotion Details
Epredia™
MS-1068-P
200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12628826
View Documents Promotion Details
Epredia™
MS-1068-P1ABX
1mg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12638826
View Documents Promotion Details
Epredia™
MS-1068-PABX
1mg/mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Keratin 15 is a type I keratin without a defined type II partner. Keratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail. Expression of keratin 15 is downregulated in some hyperproliferating situations, such as psoriasis and hypertrophic scars. Because keratinocytes in psoriasis and hypertrophic scars are activated, it is suggested that keratin 15 expression is not compatible with keratinocyte activation and the keratin 15 gene is downregulated to maintain the activated phenotype.

Host Species: Mouse

Clone: LHK15

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin 15 Ab-1
Immunofluorescence
Unconjugated
Mouse
Cytoskeletal Protein
A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
Primary
LHK15
Monoclonal
Concentrated
RUO
Bovine, Human, Mouse, Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications