missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cytokeratin 15 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunofluorescence applications with Epredia™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1068-PABX

Additional Details : Weight : 0.00180kg

 View more versions of this product

Product Code. 12638826

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Keratin 15 is a type I keratin without a defined type II partner. Keratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail. Expression of keratin 15 is downregulated in some hyperproliferating situations, such as psoriasis and hypertrophic scars. Because keratinocytes in psoriasis and hypertrophic scars are activated, it is suggested that keratin 15 expression is not compatible with keratinocyte activation and the keratin 15 gene is downregulated to maintain the activated phenotype.

Host Species: Mouse

Clone: LHK15

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin 15 Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Cytoskeletal Protein
Bovine, Human, Mouse, Rat
IgG2a κ
Immunofluorescence
LHK15
Unconjugated
A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers