Learn More
Invitrogen™ CytKick™ Max Extended Cooling Adaptor
Product Code. 16526900
Change view
Click to view available options
Quantity:
1 adapter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16526900 1 adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16526900 Supplier Invitrogen™ Supplier No. 100063482
Estimated Shipment: 11-12-2026
The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer
  • The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer.
  • This cooling block maintains samples at 2-15°C for 95-100 minutes while they are being analyzed on the instrument.
  • The cooling adaptor is brought to temperature by placing it in a refrigerator overnight or in a freezer for a minimum of two hours prior to use.
  • If this longer cooling time is not required, please see the CytKick Max Universal Cooling Adaptor, which cools samples for ∼65 minutes.
TRUSTED_SUSTAINABILITY
Specifications
Product Line CytKick
Quantity 1 adapter
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Cooling Adapter