Learn More
Cyproterone, TRC
Product Code. 30010801
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010801 100 mg 100mg
18052850 10 mg 10mg
18071050 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010801 Supplier Toronto Research Chemicals Supplier No. TRCC989090100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2098-66-0
Molecular Formula C22 H27 Cl O3
Molecular Weight (g/mol) 374.9
Synonym 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 6-chloro-1,2-dihydro-17-hydroxy-, (1β,2β)-, 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 6-chloro-1β,2β-dihydro-17-hydroxy- (8CI), Pregna-4,6-diene-3,20-dione, 6-chloro-17-hydroxy-1α,2α-methylene- (7CI), (1β,2β)-6-Chloro-1,2-dihydro-17-hydroxy-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 1,2α-Methylene-6-chloro-6-17α-hydroxyprogesterone, 6-Chloro-1,2α-methylene-4,6-pregnadien-17α-ol-3,20-dione, 6-Chloro-17-hydroxy-1α,2α-methylenepregna-4,6-diene-3,20-dione, 6-Chloro-6-dehydro-17α-hydroxy-1,2α-methyleneprogesterone, Cyproterone, SH 80881, SH 881
IUPAC Name (1S,2S,3S,5R,11R,12S,15R,16S)-15-acetyl-9-chloro-15-hydroxy-2,16-dimethylpentacyclo[9.7.0.02,8.03,5.012,16]octadeca-7,9-dien-6-one
SMILES CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C22H27ClO3/c1-11(24)22(26)7-5-14-12-9-18(23)17-10-19(25)13-8-16(13)21(17,3)15(12)4-6-20(14,22)2/h9-10,12-16,26H,4-8H2,1-3H3/t12-,13+,14-,15-,16-,20-,21-,22-/m0/s1
Formula Weight 374.1649
Percent Purity >95%
Chemical Name or Material Cyproterone
Grade HPLC