missing translation for 'onlineSavingsMsg'
Learn More

Cyproconazole, TRC

Product Code. 30137003
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137003 25 g 25g
30040866 1 g 1g
30010928 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137003 Supplier Toronto Research Chemicals Supplier No. TRCC98907025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94361-06-5
Molecular Formula C15 H18 Cl N3 O
Molecular Weight (g/mol) 291.78
Synonym α-(4-Chlorophenyl)-α-(1-cyclopropylethyl)-1H-1,2,4-triazole-1-ethanol, Alto, Alto 100, Alto 100SL, Alto 240EC, Atemi, Atemi 10 Pepite, Atemi C, Caddy, Cyproconazole, SAN 619F, SN 108266
IUPAC Name 2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(C1CC1)C(O)(Cn2cncn2)c3ccc(Cl)cc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H18ClN3O/c1-11(12-2-3-12)15(20,8-19-10-17-9-18-19)13-4-6-14(16)7-5-13/h4-7,9-12,20H,2-3,8H2,1H3
Formula Weight 291.1138
Chemical Name or Material Cyproconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.