missing translation for 'onlineSavingsMsg'
Learn More

Cyproconazole 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30324783
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324783 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324783 Supplier Dr. Ehrenstorfer Supplier No. DREA11908000AC1000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Cyproconazole
CAS 94361-06-5
Analyte or Component Names Cyproconazole
Shipping Condition Room Temperature
Molecular Formula C15 H18 Cl N3 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C15H18ClN3O/c1-11(12-2-3-12)15(20,8-19-10-17-9-18-19)13-4-6-14(16)7-5-13/h4-7,9-12,20H,2-3,8H2,1H3
SMILES CC(C1CC1)C(O)(Cn2cncn2)c3ccc(Cl)cc3
IUPAC Name 2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol
Solvent or Matrix Acetone
Molecular Weight (g/mol) 291.78
Quantity 1 mL
Formula Weight 291.1138 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.