Learn More
Tocris Bioscience™ CyPPA
GREENER_CHOICE
Product Code. 18796226
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796226 25 mg 25mg
18788296 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18796226 Supplier Tocris Bioscience™ Supplier No. 2953/25
This item is not returnable. Se returpolicy
KCa2.2 and KCa2.3 (SK) channel activator
CyPPA is an activator of small conductance Ca2+-activated K+ channels that displays selectivity for KCa2.2 (SK2) and KCa2.3 (SK3) channels (EC50 values are 5.6 and 14 μM for KCa2.3 and KCa2.2 respectively). Displays no activity at KCa2.1 (SK1) and KCa3.1 (IK) channels.
TRUSTED_SUSTAINABILITY
Tekniske data
Chemical Name or Material N-Cyclohexyl-N-[2-(3,5-dimethyl-pyrazol-1-yl)-6-methyl-4-pyrimidinamine
CAS 73029-73-9
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C16H23N5
Formula Weight Observed MW: 285.39
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.