Learn More
Cynarin, MedChemExpress
Product Code. 16473856
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473856 25 mg 25mg
16428958 10 mg 10mg
16421388 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16473856 Supplier Medchem Express Supplier No. HYN0359
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.
Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.
Specifications
Chemical Name or Material Cynarin
Physical Form Solid
CAS 30964-13-7
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C25H24O12
For Use With (Application) COVID-19-anti-virus
Synonym Cynarine
Solubility Information DMSO : ≥ 23 mg/mL (44.53 mM)
SMILES O=C([C@]1(OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.05%
Grade Research