Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Cymoxanil |
| CAS | 57966-95-7 |
| CAS Max % | ≤ 100% |
| Density | 1.310 g/cm3 (20°C) |
| Linear Formula | C7H10N4O3 |
| Beilstein | 2214018 |
| Color | Colorless |
| Melting Point | 161°C |
| Synonym | 2-cyano-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide |
| Molecular Weight (g/mol) | 198.18 g/mol |
| Show More |