Learn More
BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape
Product Code. 10094171 Shop All Brand Products
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
8 mm
10 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
108 mm
159 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
4.5 mm
6 mm
8 mm
10 mm
27 mm
Unit Size:
Each
Pack of 10
Product Code. 10094171
Brand: BRAND™ 137135
Only null left
Add to basket
Add to basket
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars, Cylindrical Shape are suitable for universal applications. Rounded, smooth surface provides effective stirring action.
  • Manufactured from PTFE
  • Rounded, smooth surface provides effective stirring action.
  • Suitable for universal applications.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 8 mm
Length (Metric) 50 mm
Material PTFE
Quantity 10/Pk.
Shape Cylinder