Learn More
Fisherbrand™ Cylindrical PTFE or Glass Stir Bars
Product Code. 14821371 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
6 mm
10 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
80 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
7 mm
8 mm
10 mm
Unit Size:
Pack of 10
Pack of 5
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
14821371 30 mm 6 mm Pack of 10
11834792 6 mm 3 mm Pack of 10
11888932 15 mm 6 mm Pack of 10
11808952 25 mm 8 mm Pack of 10
16265741 50 mm 8 mm Pack of 10
11878902 80 mm 10 mm Pack of 5
11868852 35 mm 6 mm Pack of 10
11868942 40 mm 8 mm Pack of 10
11858852 12 mm 4.5 mm Pack of 10
11587792 50 mm 6 mm Pack of 5
11868902 13 mm 6 mm Pack of 10
11828901 13 mm 8 mm Pack of 10
11848942 15 mm 4.5 mm Pack of 10
11858922 20 mm 6 mm Pack of 10
11868922 25 mm 6 mm Pack of 10
11858942 30 mm 7 mm Pack of 10
11898852 45 mm 8 mm Pack of 10
11888942 60 mm 10 mm Pack of 5
16215751 70 mm 10 mm Pack of 10
11878932 10 mm 6 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 14821371
In Stock
Add to basket
Edge
Add to basket
Alnico V magnet core encapsulated by PTFE or glass . Fisherbrand™ Cylindrical Stir Bars are autoclavable.
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 6 mm
Length (Metric) 30 mm
Quantity 5/Pk.
Shape Cylinder