Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
Product Code. 10484631
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Cone Size:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Unit Size:
Pack of 2
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Cone Size Includes Unit Size
10484631 250 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10294431 1000 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10403881 1000 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10616331 100 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10632551 100 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10303291 100 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10313101 250 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10210841 25 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10139320 50 mL 14/23 NS Pressure Equalizing Tube Pack of 2
10673891 500 mL 29/32 NS Pressure Equalizing Tube Pack of 2
10539241 50 mL 29/32 NS w/o Pressure Equalizing Tube Pack of 2
10644661 50 mL 14/23 NS w/o Pressure Equalizing Tube Pack of 2
10557321 50 mL 29/32 NS Pressure Equalizing Tube Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10484631 Supplier Lenz Laborglasinstrumente™ Supplier No. 04154914
  • ONLINE EXCLUSIVE PRICE
    127.00 EUR
    96.11 EUR / Pack of 2
    Save 30.89 EUR
    24% Off
Estimated Shipment: 23-11-2026
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Cone Size 29/32 NS
Graduated Yes
Includes w/o Pressure Equalizing Tube
Product Type Dropping Funnel