Learn More
Cyclotris(1,4-butylene Terephthalate), TRC
Product Code. 30004916
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004916 1 mg 1mg
30008111 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004916 Supplier Toronto Research Chemicals Supplier No. TRCC9889751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63440-94-8
Molecular Formula C36 H36 O12
Molecular Weight (g/mol) 660.66
Synonym 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-1(36),10,11,13(12),22,23,25(24),34,37-nonaene-2,9,14,21,26,33-hexone, 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-10,12,22,24,34,36,37,39,41-nonaene-2,9,14,21,26,33-hexone (9CI), Cyclotris(1,4-butylene Terephthalate), Polybutylene Terephthalate Cyclic Trimer, PBT Cyclic Trimer, Butylene terephthalate cyclic trimer, 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-10,12,22,24,34,36,37,39,41-nonaene-2,9,14,21,26,33-hexone, PBT Cyclic Trimer, 3,8,15,20,27,32-Hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-1(36),10,11,13(12),22,23,25(24),34,37-nonaene-2,9,14,21,26,33-hexone
IUPAC Name 3,8,15,20,27,32-hexaoxatetracyclo[32.2.2.210,13.222,25]dotetraconta-1(37),10,12,22(40),23,25(39),34(38),35,41-nonaene-2,9,14,21,26,33-hexone
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc(cc3)C(=O)OCCCCOC(=O)c4ccc1cc4
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C36H36O12/c37-31-25-7-9-27(10-8-25)33(39)45-21-3-4-23-47-35(41)29-15-17-30(18-16-29)36(42)48-24-6-5-22-46-34(40)28-13-11-26(12-14-28)32(38)44-20-2-1-19-43-31/h7-18H,1-6,19-24H2
Formula Weight 660.2207
Percent Purity >95%
Chemical Name or Material Cyclotris(1,4-butylene Terephthalate)
Grade HPLC