Learn More
Cyclotetrakis(1,4-butylene Terephthalate), TRC
Product Code. 30133108
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133108 50 mg 50mg
17918584 1 mg 1mg
17989984 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133108 Supplier Toronto Research Chemicals Supplier No. TRCC98954050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29278-72-6
Molecular Formula C48 H48 O16
Molecular Weight (g/mol) 880.8851
Synonym 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,11,12,22,23,24,34,35,36,46,48,49-dodecaene-2,9,14,21,26,33,38,45-octone, 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,12,22,24,34,36,46,48,49,51,53,55-dodecaene-2,9,14,21,26,33,38,45-octone (9CI), Terephthalic acid, tetramol. cyclic tetramethylene ester (8CI), 1,4-Butanediol, tetramol. cyclic terephthalate (8CI), Cyclotetrakis(1,4-butylene Terephthalate), Polybutylene Terephthalate Cyclic Tetramer, PBT Cyclic Tetramer, Butylene terephthalate cyclic tetramer, 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,11,12,22,23,24,34,35,36,46,48,49-dodecaene-2,9,14,21,26,33,38,45-octone, 3,8,15,20,27,32,39,44-Octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-10,12,22,24,34,36,46,48,49,51,53,55-dodecaene-2,9,14,21,26,33,38,45-octone (9CI), Terephthalic acid, tetramol. cyclic tetramethylene ester (8CI), 1,4-Butanediol, tetramol. cyclic terephthalate (8CI), Cyclotetrakis(1,4-butylene Terephthalate), PBT Cyclic Tetramer, Polybutylene Terephthalate Cyclic Tetramer (Cyclotetrakis(1,4-butylene Terephthalate))
IUPAC Name 3,8,15,20,27,32,39,44-octaoxapentacyclo[44.2.2.210,13.222,25.234,37]hexapentaconta-1(49),10,12,22,24,34(52),35,37(51),46(50),47,53,55-dodecaene-2,9,14,21,26,33,38,45-octone
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc(cc3)C(=O)OCCCCOC(=O)c4ccc(cc4)C(=O)OCCCCOC(=O)c5ccc1cc5
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C48H48O16/c49-41-33-9-11-35(12-10-33)43(51)59-27-3-4-29-61-45(53)37-17-19-39(20-18-37)47(55)63-31-7-8-32-64-48(56)40-23-21-38(22-24-40)46(54)62-30-6-5-28-60-44(52)36-15-13-34(14-16-36)42(50)58-26-2-1-25-57-41/h9-24H,1-8,25-32H2
Formula Weight 880.2942
Percent Purity >95%
Chemical Name or Material Polybutylene Terephthalate Cyclic Tetramer
Grade HPLC