Learn More
Cyclopropyldiphenylsulfonium tetrafluoroborate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 33462-81-6 |
| Molecular Formula | C15H15BF4S |
| MDL Number | MFCD00011873 |
| Quantity | 25 g |
| SMILES | F[B-](F)(F)F.[S+](C1=CC=CC=C1)(C2CC2)C3=CC=CC=C3 |
| Molecular Weight (g/mol) | 314.15 |
| Percent Purity | 95% |
| Chemical Name or Material | Cyclopropyldiphenylsulfonium tetrafluoroborate |