missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl phenyl ketone, 97%

Product Code. p-7043706
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311557 25 g 25g
11321557 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311557 Supplier Thermo Scientific Alfa Aesar Supplier No. L02218.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3481-02-5 | C10H10O | 146.19 g/mol

Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl phenyl ketone is used to prepare α-cyclopropylstyrene by the Wittig reaction in dimethyl sulfoxide. It is also used as starting reagent during the (Z)-titanium enolate formation from cyclopropyl ketones via TiCl4-n-Bu4NI-induced ring opening reaction.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3481-02-5
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001294
InChI Key PJRHFTYXYCVOSJ-UHFFFAOYSA-N
Synonym cyclopropyl phenyl ketone, cyclopropyl phenyl methanone, phenyl cyclopropyl ketone, benzoylcyclopropane, methanone, cyclopropylphenyl, cyclopropylphenylketone, ketone, cyclopropyl phenyl, cyclopropyl-phenylmethanone, acmc-2097dl, cyclopropyl-phenyl-methanone
PubChem CID 77029
IUPAC Name cyclopropyl(phenyl)methanone
SMILES C1CC1C(=O)C2=CC=CC=C2

Specifications

Density 1.06
Boiling Point 248°C
Flash Point 90°C (194°F)
Refractive Index 1.5534
Quantity 25 g
Beilstein 1860145
Solubility Information Insoluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material Cyclopropyl phenyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.