missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl 2-Fluorobenzyl Ketone, TRC

Product Code. 30009270
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009270 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009270 Supplier Toronto Research Chemicals Supplier No. TRCC98865510G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 150322-73-9
Molecular Formula C11 H11 F O
Quantity 10 g
Synonym 1-Cyclopropyl-2-(2-fluorophenyl)ethanone
InChI Formula InChI=1S/C11H11FO/c12-10-4-2-1-3-9(10)7-11(13)8-5-6-8/h1-4,8H,5-7H2
SMILES Fc1ccccc1CC(=O)C2CC2
IUPAC Name 1-cyclopropyl-2-(2-fluorophenyl)ethanone
Molecular Weight (g/mol) 178.2
Formula Weight 178.0794
Chemical Name or Material Cyclopropyl 2-Fluorobenzyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.