Learn More
Cyclopentane-d9, CDN
Product Code. 30320302
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320302 0.25 g 0.25g
30321048 0.5 g 0.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320302 Supplier CDN Supplier No. CDND37560.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cyclopentane-D9
CAS 80862-68-6
Molecular Formula C5 D9 H
Quantity 0.25 g
Synonym Cyclopentane-d9 (6 CI, 9 CI), Cyclopentane-D9
InChI Formula InChI=1 S/C5H10/c1-2-4-5-3-1/h1-5H2/i1D,2D2,3D2,4D2,5D2
SMILES [2 H]C1C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC Name 1,1,2,2,3,3,4,4,5-nonadeuteriocyclopentane
Molecular Weight (g/mol) 79.1884
Formula Weight 79.1347 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity