Learn More
Cyclopentakis(1,4-butylene Terephthalate), TRC
Product Code. 17978494
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978494 1 mg 1mg
17908884 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17978494 Supplier Toronto Research Chemicals Supplier No. TRCC9895451MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82298-33-7
Molecular Formula C60 H60 O20
Molecular Weight (g/mol) 1101.11
Synonym 3,8,15,20,27,32,39,44,51,56-Decaoxahexacyclo[56.2.2.210,13.222,25.234,37.246,49]heptaconta-10,11,12,22,23,24,34,35,36,46,47,48,58,60,61-pentadecaene-2,9,14,21,26,33,38,45,50,57-decone, 3,8,15,20,27,32,39,44,51,56-Decaoxahexacyclo[56.2.2.210,13.222,25.234,37.246,49]heptaconta-10,12,22,24,34,36,46,48,58,60,61,63,65,67,69-pentadecaene-2,9,14,21,26,33,38,45,50,57-decone (9CI), Cyclopentakis(1,4-butylene Terephthalate), Polybutylene Terephthalate Cyclic Pentamer, PBT Cyclic Pentamer, Butylene terephthalate cyclic pentamer
IUPAC Name 3,8,15,20,27,32,39,44,51,56-decaoxahexacyclo[56.2.2.210,13.222,25.234,37.246,49]heptaconta-1(61),10,12,22,24,34(66),35,37(65),46(64),47,49(63),58(62),59,67,69-pentadecaene-2,9,14,21,26,33,38,45,50,57-decone
SMILES O=C1OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCOC(=O)c3ccc(cc3)C(=O)OCCCCOC(=O)c4ccc(cc4)C(=O)OCCCCOC(=O)c5ccc(cc5)C(=O)OCCCCOC(=O)c6ccc1cc6
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C60H60O20/c61-51-41-11-13-43(14-12-41)53(63)73-33-3-4-35-75-55(65)45-19-21-47(22-20-45)57(67)77-37-7-8-39-79-59(69)49-27-29-50(30-28-49)60(70)80-40-10-9-38-78-58(68)48-25-23-46(24-26-48)56(66)76-36-6-5-34-74-54(64)44-17-15-42(16-18-44)52(62)72-32-2-1-31-71-51/h11-30H,1-10,31-40H2
Formula Weight 1100.3678
Percent Purity >95%
Chemical Name or Material Polybutylene Terephthalate Cyclic Pentamer
Grade HPLC