missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine, Free base, 98 to 99%, MP Biomedicals™

Product Code. 11462132 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462132 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462132 Supplier MP Biomedicals Supplier No. 0215074525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 108-91-8
Color Colorless
Density 0.819g/cm3 at 20°C
Assay Percent Range 98 to 99%
Odor Strong, fishy, amine odor
Molecular Formula C6H13N
MDL Number MFCD00001486
Quantity 25 mL
Synonym cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
InChI Key PAFZNILMFXTMIY-UHFFFAOYSA-N
SMILES C1CCC(CC1)N
IUPAC Name cyclohexanamine
Molecular Weight (g/mol) 99.177
PubChem CID 7965
ChEBI CHEBI:15773
Formula Weight 99.2
Physical Form Liquid
Chemical Name or Material Cyclohexylamine, Free Base
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.