Learn More
Cyclohexene, Sigma-Aldrich™
Product Code. 15898850
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898850 100 mL 100mL
15816330 500 mL 500mL
15808860 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15898850 Supplier Sigma Aldrich Supplier No. 8028240100
 Restricted Item
Estimated Shipment: 18-11-2026
Stabilized for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-83-8
Specifications
Melting Point -104°C
Density 0.81 g/cm3 at 20°C
Boiling Point 83°C/1013 hPa
MDL Number MFCD00001539
Quantity 100 mL
Synonym Cyclohexene, 1,2,3,4-Tetrahydrobenzene, Cyclohexene
Molecular Weight (g/mol) 82.14
Grade Research