missing translation for 'onlineSavingsMsg'
Learn More

Cyclofenil, TRC

Product Code. 30006490
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006490 50 mg 50mg
30005629 1 g 1g
30007446 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006490 Supplier Toronto Research Chemicals Supplier No. TRCC98783550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2624-43-3
Molecular Formula C23 H24 O4
Molecular Weight (g/mol) 364.43
Synonym Phenol, 4,4'-(cyclohexylidenemethylene)bis-, diacetate (9CI), Phenol, 4-[[4-(acetyloxy)phenyl]cyclohexylidenemethyl]-, acetate, p-Cresol, α-cyclohexylidene-α-(p-hydroxyphenyl)-, diacetate (7CI,8CI), 4,4'-(Cyclohexylidenemethylene)diphenol diacetate ester, Bis(p-acetoxyphenyl)cyclohexylidenemethane, Bis(p-hydroxyphenyl)cyclohexyldienemethane diacetate, Cyclofenil, Cyclofenyl, Cyclopenil, Cyclophenil, Cyclophenyl, F 6066, Fertodur, H 3452, ICI 48213, NSC 86464, Neoclym, Oginex, Ondogyne, Ondonid, Rehibin, Sanocrisin, Sexadieno, Sexovar, Sexovid, α-(p-Hydroxyphenyl)-α-cyclohexylidene-p-cresol diacetate, α-Cyclohexylidene-α-(p-hydroxyphenyl)-p-cresol diacetate, Phenol, 4,4'-(cyclohexylidenemethylene)bis-, 1,1'-diacetate
IUPAC Name [4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
SMILES CC(=O)Oc1ccc(cc1)C(=C2CCCCC2)c3ccc(OC(=O)C)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H24O4/c1-16(24)26-21-12-8-19(9-13-21)23(18-6-4-3-5-7-18)20-10-14-22(15-11-20)27-17(2)25/h8-15H,3-7H2,1-2H3
Formula Weight 364.1675
Chemical Name or Material Cyclofenil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.