missing translation for 'onlineSavingsMsg'
Learn More

Cyclofenil, Mikromol™

Product Code. 30051281
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051281 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051281 Supplier Mikromol Supplier No. MM3024.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cyclofenil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclofenil
CAS 2624-43-3
Analyte or Component Names Cyclofenil
Shipping Condition Room Temperature
Molecular Formula C23 H24 O4
Certifications/Compliance ISO 17025
Synonym Phenol, 4,4'-(cyclohexylidenemethylene)bis-, diacetate (9CI), Phenol, 4-[[4-(acetyloxy)phenyl]cyclohexylidenemethyl]-, acetate, 4,4'-(Cyclohexylidenemethylene)diphenol diacetate ester, Bis(p-acetoxyphenyl)cyclohexylidenemethane, Bis(p-hydroxyphenyl)cyclohexyldienemethane diacetate, Cyclofenil, Cyclofenyl, Cyclopenil, Cyclophenil, Cyclophenyl, F 6066, Fertodur, H 3452, ICI 48213, NSC 86464, Neoclym, Oginex, Ondogyne, Ondonid, Rehibin, Sanocrisin, Sexadieno, Sexovar, Sexovid, Phenol, 4,4'-(cyclohexylidenemethylene)bis-, 1,1'-diacetate, p-Cresol, alpha-cyclohexylidene-alpha-(p-hydroxyphenyl)-, diacetate (7CI,8CI), alpha-(p-Hydroxyphenyl)-alpha-cyclohexylidene-p-cresol diacetate, alpha-Cyclohexylidene-alpha-(p-hydroxyphenyl)-p-cresol diacetate
InChI Formula InChI=1S/C23H24O4/c1-16(24)26-21-12-8-19(9-13-21)23(18-6-4-3-5-7-18)20-10-14-22(15-11-20)27-17(2)25/h8-15H,3-7H2,1-2H3
SMILES CC(=O)Oc1ccc(cc1)C(=C2CCCCC2)c3ccc(OC(=O)C)cc3
IUPAC Name [4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
Molecular Weight (g/mol) 364.43
Quantity 250 mg
Formula Weight 364.1675 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.