Learn More
Epredia™ Lab Vision™ Cyclin D1/Bcl-1 Ab-1 Mouse Monoclonal Antibody, 1mL; 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications.
Brand: Epredia™ MS-210-P0
Description
Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators and is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL)Host Species: Mouse
Clone: DCS-6
Isotype: IgG2a/κ
Species Reactivity: Human, Monkey, Mouse, Rat and Dog.
Epitope: Not determined
Immunogen: Human recombinant full-length cyclin D1 protein
Molecular Weight: 36 kDa
Positive Control: MAD109 cells; Mantle cell lymphomas and breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Biological Blockade
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| Cyclin D1/Bcl-1 Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Cell Cycle | |
| Canine, Human, Mouse, Non-human Primate, Rat | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| DCS-6 | |
| Unconjugated | |
| Human recombinant full length cyclin D1 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.