missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cyclin D1/Bcl-1 Ab-1 Mouse Monoclonal Antibody, 1mL; 200μg/mL, biotinylated, BSA and azide

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications.

177.00 EUR

Specifications

Antigen Cyclin D1/Bcl-1 Ab-1
Clone DCS-6
Concentration 200μg/mL
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12630557
View Documents
Epredia™
MS-210-P0
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators and is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL)

Host Species: Mouse

Clone: DCS-6

Isotype: IgG2a/κ

Species Reactivity: Human, Monkey, Mouse, Rat and Dog.

Epitope: Not determined

Immunogen: Human recombinant full-length cyclin D1 protein

Molecular Weight: 36 kDa

Positive Control: MAD109 cells; Mantle cell lymphomas and breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cyclin D1/Bcl-1 Ab-1
200μg/mL
Monoclonal
Concentrated
RUO
Canine, Human, Mouse, Non-human Primate, Rat
IgG2a κ
DCS-6
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Cell Cycle
Human recombinant full length cyclin D1 protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.