missing translation for 'onlineSavingsMsg'
Learn More

Cyclen, MedChemExpress

Product Code. 16439326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439326 Supplier Medchem Express Supplier No. HYW007656
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Cyclen is the aza analogue of crown ether, used as a precursor for MRI contrast agents, and is an intermediate for the preparation of effective macrocyclic chelates.

Cyclen is the aza analogue of crown ether, used as a precursor for MRI contrast agents, and is an intermediate for the preparation of effective macrocyclic chelates.

Specifications

Chemical Name or Material Cyclen
CAS 294-90-6
Color White
Purity Grade Notes Research
Molecular Formula C8H20N4
Quantity 100 mg
Solubility Information DMSO : 1.85 mg/mL (10.74 mM; ultrasonic and warming and heat to 60°C)
SMILES C1CNCCNCCNCCN1
Molecular Weight (g/mol) 172.27
Formula Weight 172.27
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.