Learn More
(Cyanomethyl)urea, TRC
Product Code. 30004733
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004733 50 mg 500mg
17993325 250 mg 250mg
17931515 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004733 Supplier Toronto Research Chemicals Supplier No. TRCC27350050MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5962-07-2
Molecular Formula C3 H5 N3 O
Molecular Weight (g/mol) 99.091
Synonym Urea, (cyanomethyl)- (8CI,9CI), N-(Cyanomethyl)urea, (Cyanomethyl)urea, Acetonitrile, [(aminocarbonyl)amino]-, Carbamyl glycinonitrile
IUPAC Name cyanomethylurea
SMILES NC(=O)NCC#N
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C3H5N3O/c4-1-2-6-3(5)7/h2H2,(H3,5,6,7)
Formula Weight 99.043
Percent Purity >95%
Chemical Name or Material Hydantoic Acid Nitrile (Cyanomethylurea)
Grade HPLC