missing translation for 'onlineSavingsMsg'
Learn More

Cyanomelamine, TRC

Product Code. 30136741
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136741 25 mg 25mg
30134310 100 mg 100mg
17972625 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136741 Supplier Toronto Research Chemicals Supplier No. TRCC98453025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3496-98-8
Molecular Formula C4 H5 N7
Molecular Weight (g/mol) 151.13
Synonym 2-(Cyanoamino)-4,6-diamino-sym-triazine, 4,6-Diamino-s-triazine-2-carbamonitrile, (4,6-Diamino-1,3,5-triazin-2-yl)cyanamide, N-(4,6-diamino-1,3,5-triazin-2-yl)cyanamide
IUPAC Name (4,6-diamino-1,3,5-triazin-2-yl)cyanamide
SMILES Nc1nc(N)nc(NC#N)n1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C4H5N7/c5-1-8-4-10-2(6)9-3(7)11-4/h(H5,6,7,8,9,10,11)
Formula Weight 151.0606
Chemical Name or Material Cyanomelamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.