missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen Bromide, Sigma-Aldrich™

Product Code. 15884491
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884491 250 g 250g
15874491 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884491 Supplier Sigma Aldrich Supplier No. 8201940250
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-68-3

Specifications

Melting Point 52°C
Density 2.015 g/cm3 at 20°C
Boiling Point 61°C to 62°C/1013 hPa
MDL Number MFCD00011597
Quantity 250 g
Synonym Cyanogen bromide, Cyanogen bromide
Molecular Weight (g/mol) 105.92
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.