missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide-13C, TRC

Product Code. 30136152
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136152 500 mg 500mg
30003480 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136152 Supplier Toronto Research Chemicals Supplier No. TRCC952502500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21420-35-9
Molecular Formula 13C H2 N2
Molecular Weight (g/mol) 43.03
IUPAC Name aminoformonitrile
SMILES N[13C]#N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/CH2N2/c2-1-3/h2H2/i1+1
Formula Weight 43.0252
Chemical Name or Material Cyanamide-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.