Learn More
MedChemExpress CY5-YNE
Click to view available options
:
5 mg
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 693.87 g/mol |
| Solubility | DMSO : ≥ 43 mg/mL (61.97 mM) |
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 1345823-20-2 |
| Synonym | Sulfo-Cyanine5-alkyne |