Learn More
Cytiva Cy™5 Amidite
Product Code. 11520654 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11520654 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11520654 Supplier Cytiva Supplier No. 28904249
  • ONLINE EXCLUSIVE PRICE
    497.00 EUR
    406.76 EUR / 100mg
    Save 90.24 EUR
    18% Off
 Restricted Item
Estimated Shipment: 13-11-2026
For CyDye™ incorporation at any position in an oligonucleotide during synthesis
CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.
  • Wide selection of CyDye™ fluors for protein labeling for use in ELISA & immunoprecipitation
  • Labeling strategy based on protein characteristics
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Streptavidin-conjugated CyDye™ fluors for use in Flow-cytometry and In situ hybridization
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
  • DNA synthesis reagent
Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye™ fluors enables multiplexing without crosstalk. In addition, the CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham™ CyDye™ fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.
TRUSTED_SUSTAINABILITY
Specifications
Color Dark blue
Content And Storage Store in original container protected from direct sunlight in dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use
For Use With (Application) Prepare sequencing primers for use on ALFexpress, industrial applications such as analytical reagent and research
Product Type Flourescet dye
Quantity 100 mg
Sample Volume 100 mg
compliance-icons compliance-icons
Product Identifier
  • Cy 5 Amidite, 100 mg
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .