Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
Product Code. 12872003
Change view
Click to view available options
Type:
Disposable Cuvette
Unit Size:
Pack of 51
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12872003 Disposable Cuvette Pack of 51
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12872003 Supplier Reichert™ Supplier No. 13102140
  • ONLINE EXCLUSIVE PRICE
    116.53 EUR
    110.21 EUR / Pack of 51
    Save 6.32 EUR
    5% Off
Estimated Shipment: 14-01-2027
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
  • For use with Unistat Bilirubinometer
  • Capacity: 20μL
Specifications
Type Disposable Cuvette
Sample Type Undiluted Serum
For Use With (Equipment) Reichert Unistat™ Bilirubinometer
Capacity (Metric) 20 μL