Learn More
Sampling Systems™ Cup Liquid Sampler
Product Code. 11930018 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
100 mL
500 mL
Length (Metric):
600 mm
1000 mm
1500 mm
Diameter (Metric) Outer:
55 mm
80 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) Outer Unit Size
11930018 100 mL 1000 mm 55 mm Each
11920018 100 mL 600 mm 55 mm Each
11935437 500 mL 1000 mm 80 mm Each
11945437 500 mL 1500 mm 80 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11930018 Supplier Sampling Systems™ Supplier No. 5030A1000
Estimated Shipment: 14-12-2026
Constructed of stainless steel and for sampling liquids and suspensions. Sampling Systems™ Cup Liquid Sampler enables discrete samples to be easily taken from a height within the product.
  • 316L Stainless Steel Construction
  • Single point liquid sampler
Specifications
Product Type Liquid Sampler
Material Stainless Steel
Capacity (Metric) 100 mL
Diameter (Metric) Outer 55 mm
Length (Metric) 1000 mm