Learn More
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck
Product Code. 11562283 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11562283 2000 mL Each
11532283 250 mL Each
11542283 500 mL Each
11552283 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11562283
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
Culture your tissues using the Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck. It is designed for consistency, cell health and reproducability with surface and ancillary options you're looking for in a issue culture flask.
  • Fluted neck
  • For adherent or suspension cells
  • 1/3 turn cap enables one-handed operation
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Neck Style Fluted
Shape Conical
Diameter (Metric) Outer 165 mm