Learn More
Thermo Scientific™ Culti-Loops™ Lactobacillus acidophilus ATCC™ 314™
Product Code. 12958988
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12958988 5 Loops/Pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12958988 Supplier Thermo Scientific™ Supplier No. R4603050
  • ONLINE EXCLUSIVE PRICE
    240.00 EUR
    209.68 EUR / Pack of 5
    Save 30.32 EUR
    12% Off
 Restricted Item
Estimated Shipment: 08-12-2026
If you would like to order, a signed ATCC agreement is required.
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 314TM*
Description Lactobacillus acidophilus
Detects Lactobacillus acidophilus
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations WDCM00031, [43]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.