missing translation for 'onlineSavingsMsg'
Learn More

CUDC-427, MedChemExpress

Product Code. p-200097593
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469397 5 mg 5mg
16466858 10 mg 10mg
16442927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16469397 Supplier Medchem Express Supplier No. HY15835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CUDC-427 is a potent second-generation pan-selective IAP antagonist, used for treatment of various cancers.

CUDC-427 is a potent second-generation pan-selective IAP antagonist, used for treatment of various cancers.

Specifications

Chemical Name or Material CUDC-427
CAS 1446182-94-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H36N6O4S
Quantity 5 mg
Synonym GDC-0917
Solubility Information DMSO : ≥ 100 mg/mL (177.09 mM)
SMILES O=C([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)NC3=C(C4=CC=CC=C4)N=C(C5=NC=CO5)S3
Molecular Weight (g/mol) 564.7
Formula Weight 564.7
Percent Purity 96.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.